Assay Details
Target Gene Details
Entrez Gene ID: | 9330 |
Gene Name: | general transcription factor IIIC subunit 3 |
Gene Aliases: |
TFIIIC102, TFIIICgamma, TFiiiC2-102 |
Location: |
Chr.2:196763032-196799768 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C3 | NM_012086.4 | 15 | 2293 | NP_036218.1 |
| XM_005246965.4 | 15 | 2297 | XP_005247022.1 | |
| AF133123.1 | 15 | 2229 | AAD41475.1 | |
| AK025560.1 | 15 | 2207 | ||
| AK027050.1 | 9 | 1244 | BAB15638.1 | |
| AK314718.1 | 15 | 2234 | ||
| AL137453.1 | 12 | 1635 | CAB70745.1 | |
| BC015995.1 | 7 | 992 | AAH15995.1 | |
| BC043347.1 | 15 | 2148 | AAH43347.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575323 | Chr.2:196674745 - 197095661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map