Assay Details
Target Gene Details
Entrez Gene ID: | 54821 |
Gene Name: | ERCC excision repair 6 like, spindle assembly checkpoint helicase |
Gene Aliases: |
PICH, RAD26L |
Location: |
Chr.X:72204657-72239008 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC6L | NM_017669.2 | 2 | 3978 | NP_060139.2 |
| AK000112.1 | 1 | 2075 | BAA90952.1 | |
| AK056494.1 | 3 | 4115 | ||
| AK074719.1 | 1 | 3452 | BAC11160.1 | |
| AW015865.1 | 1 | 247 | ||
| AY121802.1 | 1 | 2075 | AAM82750.1 | |
| BC008808.2 | 1 | 1492 | ||
| BC111486.1 | 2 | 3901 | AAI11487.1 |
Target Gene Details
Entrez Gene ID: | 5303 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 4 |
Gene Aliases: |
EPVH, PAR14, PAR17 |
Location: |
Chr.X:72181676-72263964 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIN4 | NM_001170747.1 | NP_001164218.1 | ||
| AK127605.1 |
More Information
Set Membership: |
|

Genomic Map