Assay Details
Target Gene Details
Entrez Gene ID: | 353322 |
Gene Name: | ankyrin repeat domain 37 |
Gene Aliases: |
Lrp2bp |
Location: |
Chr.4:185396686-185400245 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD37 | NM_181726.3 | NP_859077.1 | ||
| XM_017008176.1 | XP_016863665.1 | |||
| AK303243.1 | ||||
| AY296056.1 | AAP50252.1 | |||
| BC088376.1 | AAH88376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033523 | Chr.4:185394656 - 185473803 on Build GRCh38 | Gain |
|
| esv3603492 | Chr.4:185370615 - 185448567 on Build GRCh38 | Gain |
|
| esv3603493 | Chr.4:185372047 - 185422056 on Build GRCh38 | Gain |
|
| nsv428456 | Chr.4:185328666 - 185492417 on Build GRCh38 | Gain+Loss |
|
| esv2759309 | Chr.4:185391971 - 185553542 on Build GRCh38 | Loss |
|

Genomic Map