Assay Details
Target Gene Details
Entrez Gene ID: | 285203 |
Gene Name: | EGF domain specific O-linked N-acetylglucosamine transferase |
Gene Aliases: |
AER61, AOS4, C3orf64, EOGT1 |
Location: |
Chr.3:68975212-69013961 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EOGT | NM_001278689.1 | 18 | 3546 | NP_001265618.1 |
| NM_173654.2 | 15 | 3294 | NP_775925.1 | |
| NR_103826.1 | 16 | 3335 | ||
| XM_005264743.2 | 18 | 3760 | XP_005264800.1 | |
| XM_011533599.1 | 19 | 3814 | XP_011531901.1 | |
| XM_011533600.1 | 19 | 3667 | XP_011531902.1 | |
| XM_011533602.1 | 19 | 3579 | XP_011531904.1 | |
| XM_011533603.1 | 19 | 3811 | XP_011531905.1 | |
| XM_017006204.1 | 19 | 3664 | XP_016861693.1 | |
| XM_017006205.1 | 18 | 3525 | XP_016861694.1 | |
| XM_017006206.1 | 18 | 3421 | XP_016861695.1 | |
| XM_017006207.1 | 11 | 2260 | XP_016861696.1 | |
| XM_017006209.1 | 9 | 2133 | XP_016861698.1 | |
| AK023140.1 | 1 | 1141 | ||
| AK091089.1 | 9 | 2524 | ||
| AK123213.1 | 6 | 1858 | ||
| AK126187.1 | 14 | 2618 | BAC86479.1 | |
| AL833467.1 | 4 | 1700 | ||
| BC028935.1 | 1 | 827 | AAH28935.1 | |
| BC060887.1 | 15 | 3023 | AAH60887.1 | |
| BX640821.1 | 14 | 2667 | CAE45897.2 | |
| BX648818.1 | 18 | 3321 |
More Information
Additional Information:
For this assay, SNP(s) [rs115176538,rs80297124] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map