Assay Details
Target Gene Details
Entrez Gene ID: | 4054 |
Gene Name: | latent transforming growth factor beta binding protein 3 |
Gene Aliases: |
DASS, LTBP-3, LTBP2, STHAG6, pp6425 |
Location: |
Chr.11:65538559-65558388 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTBP3 | NM_001130144.2 | 28 | 4386 | NP_001123616.1 |
| NM_001164266.1 | 27 | 4241 | NP_001157738.1 | |
| NM_021070.4 | 27 | 4245 | NP_066548.2 | |
| XM_011545032.2 | 28 | 4568 | XP_011543334.1 | |
| XM_011545033.2 | 27 | 4426 | XP_011543335.1 | |
| AF135960.3 | 27 | 3976 | AAF62352.3 | |
| AF318354.1 | 26 | 3417 | AAL55861.1 | |
| AK024477.1 | ||||
| AK090750.1 | ||||
| AK097212.1 | ||||
| AK124524.1 | ||||
| AK126126.1 | ||||
| AL117551.1 | CAB55988.1 | |||
| BC008761.2 | AAH08761.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555212 | Chr.11:65535422 - 65540385 on Build GRCh38 | Loss |
|
| esv2671903 | Chr.11:65538274 - 65540821 on Build GRCh38 | Deletion |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map