Assay Details
Target Gene Details
Entrez Gene ID: | 55679 |
Gene Name: | LIM zinc finger domain containing 2 |
Gene Aliases: |
LGMD2W, PINCH-2, PINCH2 |
Location: |
Chr.2:127638421-127681786 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4068n100 | Chr.2:127537744 - 127672201 on Build GRCh38 | Gain |
|
| nsv517532 | Chr.2:127630362 - 127647285 on Build GRCh38 | Loss |
|
| esv3893304 | Chr.2:127634972 - 127643416 on Build GRCh38 | Loss |
|
| nsv508864 | Chr.2:127585891 - 127684658 on Build GRCh38 | Insertion |
|
| nsv518085 | Chr.2:127576838 - 127647285 on Build GRCh38 | Loss |
|
| nsv953155 | Chr.2:127617926 - 127665326 on Build GRCh38 | Deletion |
|
| nsv470482 | Chr.2:127593233 - 127707973 on Build GRCh38 | Loss |
|
| nsv583037 | Chr.2:127576895 - 127643516 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map