Assay Details
Target Gene Details
Entrez Gene ID: | 10732 |
Gene Name: | transcription factor like 5 |
Gene Aliases: |
CHA, E2BP-1, Figlb, bHLHe82 |
Location: |
Chr.20:62841014-62861763 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCFL5 | NM_006602.3 | 6 | 1812 | NP_006593.2 |
| XM_005260185.3 | 6 | 1716 | XP_005260242.1 | |
| AB012124.1 | 6 | 1673 | BAA36557.1 | |
| AF070992.1 | 6 | 1650 | AAD53986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834027 | Chr.20:62707655 - 62874914 on Build GRCh38 | Loss |
|
| nsv828794 | Chr.20:62832958 - 62880035 on Build GRCh38 | Loss |
|
| nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
|
| nsv428380 | Chr.20:62632018 - 62935383 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map