Assay Details
Target Gene Details
Entrez Gene ID: | 51076 |
Gene Name: | cutC copper transporter |
Gene Aliases: |
CGI-32 |
Location: |
Chr.10:99732201-99756137 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUTC | NM_015960.2 | NP_057044.2 | ||
| AF132966.1 | AAD27741.1 | |||
| AK001142.1 | ||||
| AK302507.1 | ||||
| AK314687.1 | ||||
| BC021105.1 | AAH21105.1 | |||
| BC028948.1 | AAH28948.1 |
More Information
Set Membership: |
|

Genomic Map