Assay Details
Target Gene Details
Entrez Gene ID: | 164045 |
Gene Name: | HFM1, ATP dependent DNA helicase homolog |
Gene Aliases: |
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase |
Location: |
Chr.1:91260766-91408008 on Build GRCh38 |
Assay Gene Location: | Within Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HFM1 | NM_001017975.4 | 34 | 3828 | NP_001017975.4 |
| XM_011540849.1 | 34 | 3911 | XP_011539151.1 | |
| XM_011540850.2 | 34 | 3840 | XP_011539152.1 | |
| XM_011540851.1 | 34 | 3795 | XP_011539153.1 | |
| XM_011540852.2 | 34 | 3777 | XP_011539154.1 | |
| XM_011540855.1 | 32 | 3633 | XP_011539157.1 | |
| XM_011540857.1 | 32 | 3392 | XP_011539159.1 | |
| XM_011540859.2 | 31 | 3361 | XP_011539161.1 | |
| XM_017000490.1 | 33 | 3691 | XP_016855979.1 | |
| XM_017000491.1 | 33 | 3640 | XP_016855980.1 | |
| XM_017000492.1 | 28 | 2875 | XP_016855981.1 | |
| XM_017000493.1 | 22 | 2079 | XP_016855982.1 | |
| AB044787.1 | 11 | 1116 | ||
| AB074274.1 | 6 | 579 | ||
| AB204867.1 | 34 | 3829 | ||
| AK094079.1 | 17 | 1655 | BAC04281.1 | |
| AK294754.1 | 28 | 2846 | ||
| AK302582.1 | 30 | 2996 | ||
| BC144172.1 | 19 | 1774 | ||
| BX649037.1 | 15 | 1364 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map