Assay Details
Target Gene Details
Entrez Gene ID: | 9236 |
Gene Name: | cell cycle progression 1 |
Gene Aliases: |
CPR8 |
Location: |
Chr.15:55355223-55408510 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCPG1 | NM_001204450.1 | 7 | 1053 | NP_001191379.1 |
| NM_001204451.1 | 7 | 1053 | NP_001191380.1 | |
| NM_004748.4 | 7 | 1293 | NP_004739.3 | |
| NM_020739.3 | 7 | 1053 | NP_065790.2 | |
| AB033080.2 | 7 | 838 | ||
| AF212228.1 | 7 | 969 | AAK14914.1 | |
| AK022459.1 | 7 | 849 | BAB14042.1 | |
| AK292795.1 | 7 | 802 | ||
| BC015203.1 | 7 | 889 | ||
| BC034914.1 | 2 | 1211 | AAH34914.1 | |
| BC039871.1 | 7 | 807 | AAH39871.1 |
Target Gene Details
Entrez Gene ID: | 100533483 |
Gene Name: | DYX1C1-CCPG1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.15:55355223-55498584 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYX1C1-CCPG1 | NR_037923.1 | 14 | 2171 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| nsv520799 | Chr.15:55353463 - 55468324 on Build GRCh38 | Loss |
|
| nsv1043789 | Chr.15:55108674 - 55800256 on Build GRCh38 | Gain |
|
| nsv569523 | Chr.15:55263404 - 55544506 on Build GRCh38 | Gain |
|
| esv3569598 | Chr.15:55270429 - 55410270 on Build GRCh38 | Gain |
|
| esv3636557 | Chr.15:55363174 - 55366270 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map