Assay Details
Target Gene Details
Entrez Gene ID: | 56897 |
Gene Name: | Werner helicase interacting protein 1 |
Gene Aliases: |
WHIP, bA420G6.2 |
Location: |
Chr.6:2765341-2785745 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WRNIP1 | NM_020135.2 | 4 | 1506 | NP_064520.2 |
| NM_130395.2 | 4 | 1431 | NP_569079.1 | |
| AB056152.1 | 4 | 1507 | BAB60709.1 | |
| AB209723.1 | 4 | 1259 | BAD92960.1 | |
| AF218313.1 | 4 | 893 | AAF80563.1 | |
| AK026179.1 | 3 | 411 | BAB15383.1 | |
| AK094823.1 | 4 | 1466 | ||
| AK223593.1 | 4 | 1440 | BAD97313.1 | |
| AK315047.1 | 4 | 1598 | ||
| AK315471.1 | 4 | 1400 | ||
| BC018923.2 | 4 | 1392 | AAH18923.1 |
More Information
Set Membership: |
|

Genomic Map