Assay Details
Target Gene Details
Entrez Gene ID: | 286207 |
Gene Name: | cilia and flagella associated protein 157 |
Gene Aliases: |
C9orf117 |
Location: |
Chr.9:127706261-127716002 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP157 | NM_001012502.2 | NP_001012520.2 | ||
| XM_006717064.3 | XP_006717127.2 | |||
| XM_011518559.2 | XP_011516861.2 | |||
| XM_017014629.1 | XP_016870118.1 | |||
| XM_017014630.1 | XP_016870119.1 | |||
| AK094948.1 | ||||
| AK126437.1 | ||||
| BC133027.1 | ||||
| BC141809.1 |
Target Gene Details
Entrez Gene ID: | 138428 |
Gene Name: | peptidyl-tRNA hydrolase 1 homolog |
Gene Aliases: |
C9orf115, PTH1 |
Location: |
Chr.9:127695024-127716714 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRH1 | XM_017014277.1 | XP_016869766.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map