Assay Details
Target Gene Details
Entrez Gene ID: | 9354 |
Gene Name: | ubiquitination factor E4A |
Gene Aliases: |
E4, UBOX2, UFD2 |
Location: |
Chr.11:118359581-118399211 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE4A | NM_001204077.1 | NP_001191006.1 | ||
| NM_004788.3 | NP_004779.2 | |||
| AK296369.1 | ||||
| BC030973.1 | ||||
| BC052643.1 | AAH52643.1 | |||
| BC062695.1 | AAH62695.1 | |||
| BC111417.1 | ||||
| BC112367.1 | AAI12368.1 | |||
| BU167020.1 | ||||
| D50916.1 |
More Information
Set Membership: |
|

Genomic Map