Assay Details
Target Gene Details
Entrez Gene ID: | 340562 |
Gene Name: | spermidine/spermine N1-acetyl transferase-like 1 |
Gene Aliases: |
- |
Location: |
Chr.X:85092286-85116229 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATL1 | NM_001012980.2 | NP_001012998.2 | ||
| XM_011530940.2 | XP_011529242.1 | |||
| BC043215.1 | AAH43215.2 | |||
| BC126401.1 |
More Information
Set Membership: |
|

Genomic Map