Assay Details
Target Gene Details
Entrez Gene ID: | 64976 |
Gene Name: | mitochondrial ribosomal protein L40 |
Gene Aliases: |
L40mt, MRP-L22, MRP-L40, MRPL22, NLVCF, URIM |
Location: |
Chr.22:19431902-19436078 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL40 | NM_001318151.1 | NP_001305080.1 | ||
| NM_001318152.1 | NP_001305081.1 | |||
| NM_003776.3 | NP_003767.2 | |||
| AF034091.1 | AAC70904.1 | |||
| AJ295637.1 | CAC00535.1 | |||
| AK057708.1 | 1 | 2148 | ||
| AK123768.1 | ||||
| BC009707.1 | AAH09707.1 | |||
| BM974354.1 | ||||
| BP309678.1 | ||||
| BX382916.2 | ||||
| CN409565.1 | ||||
| CR456532.1 | CAG30418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834124 | Chr.22:19263316 - 19451297 on Build GRCh38 | Loss |
|
| esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain |
|
| nsv834127 | Chr.22:19414564 - 19656457 on Build GRCh38 | Loss |
|
| nsv588216 | Chr.22:19086099 - 19839117 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map