Assay Details
Target Gene Details
Entrez Gene ID: | 55768 |
Gene Name: | N-glycanase 1 |
Gene Aliases: |
CDDG, CDG1V, PNG1, PNGase |
Location: |
Chr.3:25718944-25790039 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGLY1 | NM_001145293.1 | 12 | 2108 | NP_001138765.1 |
| NM_001145294.1 | 12 | 2102 | NP_001138766.1 | |
| NM_001145295.1 | 11 | 1984 | NP_001138767.1 | |
| NM_018297.3 | 12 | 2162 | NP_060767.2 | |
| XM_005265316.1 | 11 | 1942 | XP_005265373.1 | |
| XM_005265317.1 | 10 | 1764 | XP_005265374.1 | |
| XM_011533944.1 | 12 | 2058 | XP_011532246.1 | |
| AB114414.1 | ||||
| AB209549.1 | 4 | 3104 | BAD92786.1 | |
| AF250924.2 | 12 | 2095 | AAF74720.2 | |
| AK001867.1 | 10 | 1763 | BAA91950.1 | |
| AK296047.1 | ||||
| AK310473.1 | 11 | 2957 | ||
| AL832748.1 | 3 | 1861 | ||
| BC000963.2 | 12 | 1991 | AAH00963.1 | |
| BC007226.1 | 12 | 2095 | AAH07226.1 | |
| BC017220.2 | 11 | 1911 | AAH17220.1 | |
| BQ017148.1 | 1 | 445 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893676 | Chr.3:24984204 - 26577498 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map