Assay Details
Target Gene Details
Entrez Gene ID: | 1602 |
Gene Name: | dachshund family transcription factor 1 |
Gene Aliases: |
DACH |
Location: |
Chr.13:71437966-71868068 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACH1 | NM_004392.6 | 7 | 2142 | NP_004383.4 |
| NM_080759.5 | 11 | 2748 | NP_542937.3 | |
| NM_080760.5 | 8 | 2304 | NP_542938.3 | |
| XM_011534939.2 | 12 | 3780 | XP_011533241.1 | |
| XM_011534942.2 | 10 | 1614 | XP_011533244.1 | |
| XM_017020396.1 | 11 | 1774 | XP_016875885.1 | |
| AF102546.1 | 11 | 2748 | AAF01351.1 | |
| AF356492.1 | 11 | 2748 | AAL08487.1 | |
| AJ005670.1 | 11 | 2001 | CAA06666.1 | |
| BQ440916.1 | 1 | 231 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471153 | Chr.13:71435470 - 71473964 on Build GRCh38 | Loss |
|
| nsv1038176 | Chr.13:70898018 - 71772254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map