Assay Details
Target Gene Details
Entrez Gene ID: | 80755 |
Gene Name: | alanyl-tRNA synthetase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:42950526-42964498 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AARSD1 | NM_001261434.1 | NP_001248363.1 | ||
| AK309477.1 | ||||
| AL515977.3 | ||||
| BP368900.1 | ||||
| CA951145.1 |
Target Gene Details
Entrez Gene ID: | 100885850 |
Gene Name: | PTGES3L-AARSD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:42950526-42980528 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES3L-AARSD1 | NM_001136042.2 | NP_001129514.2 | ||
| NM_025267.3 | NP_079543.1 | |||
| AF039235.1 | ||||
| AK094084.1 | ||||
| AK295447.1 | ||||
| BC004172.2 | AAH04172.1 | |||
| BC019324.1 | AAH19324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2057 | Chr.17:42946856 - 42980330 on Build GRCh38 | Insertion |
|
| nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
|
| nsv575052 | Chr.17:42907778 - 43019464 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map