Assay Details
Target Gene Details
Entrez Gene ID: | 25874 |
Gene Name: | mitochondrial pyruvate carrier 2 |
Gene Aliases: |
BRP44 |
Location: |
Chr.1:167916675-167937069 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPC2 | NM_001143674.3 | 6 | 842 | NP_001137146.1 |
| NM_015415.3 | 6 | 776 | NP_056230.1 | |
| NR_026550.2 | 5 | 1014 | ||
| XM_006711266.3 | XP_006711329.1 | |||
| AK290126.1 | ||||
| AL035304.1 | CAA22909.1 | |||
| AL110297.1 | CAB53738.1 | |||
| AL554033.3 | ||||
| BC104157.1 | AAI04158.1 | |||
| BC104158.1 | AAI04159.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32660 | Chr.1:167825845 - 168220986 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map