Assay Details
Target Gene Details
Entrez Gene ID: | 153443 |
Gene Name: | serum response factor binding protein 1 |
Gene Aliases: |
BUD22, P49, Rlb1, STRAP, p49/STRAP |
Location: |
Chr.5:121961917-122075624 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRFBP1 | NM_152546.2 | NP_689759.2 | ||
| XM_017009111.1 | XP_016864600.1 | |||
| AK058015.1 | BAB71631.1 | |||
| AY611630.2 | ||||
| BC031222.1 | AAH31222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599647 | Chr.5:121876230 - 121966379 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map