Assay Details
Target Gene Details
Entrez Gene ID: | 5801 |
Gene Name: | protein tyrosine phosphatase, receptor type R |
Gene Aliases: |
EC-PTP, PCPTP1, PTP-SL, PTPBR7, PTPRQ |
Location: |
Chr.12:70638073-70920804 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPRR | NM_001207015.1 | 13 | 2419 | NP_001193944.1 |
NM_001207016.1 | 11 | 2209 | NP_001193945.1 | |
NM_002849.3 | 14 | 3075 | NP_002840.2 | |
NM_130846.2 | 10 | 2341 | NP_570897.2 | |
NR_073474.1 | 11 | 2565 | ||
XM_011538615.2 | 14 | 2671 | XP_011536917.1 | |
AK091647.1 | 5 | 1314 | ||
BC110900.1 | 10 | 2038 | AAI10901.1 | |
BX571751.1 | 10 | 2394 | CAE11876.1 | |
CA435261.1 | 1 | 442 | ||
CR749836.1 | 5 | 1474 | CAH18692.1 | |
D64053.1 | 14 | 3077 | BAA10930.1 | |
U42361.1 | 14 | 2835 | AAD09447.1 | |
U77917.1 | 10 | 2318 | AAB54007.1 | |
X82635.1 | 10 | 2299 | CAA57957.1 | |
Z79693.1 | 14 | 2746 | CAB01957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32743 | Chr.12:70481471 - 70719432 on Build GRCh38 | Gain |
![]() ![]() |
nsv509472 | Chr.12:70606856 - 70653543 on Build GRCh38 | Insertion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
