Assay Details
Target Gene Details
Entrez Gene ID: | 3422 |
Gene Name: | isopentenyl-diphosphate delta isomerase 1 |
Gene Aliases: |
IPP1, IPPI1 |
Location: |
Chr.10:1039419-1056716 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDI1 | NM_001317955.1 | 6 | 1556 | NP_001304884.1 |
| NM_001317956.1 | 5 | 1690 | NP_001304885.1 | |
| NM_001317957.1 | 4 | 1517 | NP_001304886.1 | |
| NM_004508.3 | 5 | 1573 | NP_004499.2 | |
| NR_134300.1 | 4 | 1305 | ||
| NR_134301.1 | 4 | 1400 | ||
| XM_011519457.1 | 5 | 2420 | XP_011517759.1 | |
| XM_017016191.1 | 4 | 2247 | XP_016871680.1 | |
| AF271720.1 | 4 | 1058 | AAK49435.1 | |
| AK222875.1 | BAD96595.1 | |||
| AK303669.1 | ||||
| AK311110.1 | 4 | 949 | ||
| BC005247.2 | 4 | 1071 | AAH05247.2 | |
| BC006999.2 | 4 | 1071 | AAH06999.2 | |
| BC019227.2 | 4 | 1071 | AAH19227.2 | |
| BC022418.2 | 4 | 1071 | AAH22418.2 | |
| BC025375.2 | 4 | 1071 | AAH25375.2 | |
| BC057827.1 | 6 | 1544 | AAH57827.1 | |
| BC107893.1 | 5 | 1097 | AAI07894.1 | |
| BX537663.1 | 4 | 4117 | ||
| BX648472.1 | 4 | 2036 | ||
| X17025.1 | 5 | 1092 | CAA34890.1 |
Target Gene Details
Entrez Gene ID: | 55853 |
Gene Name: | IDI2 antisense RNA 1 |
Gene Aliases: |
C10orf110, HT009, IDI2-AS |
Location: |
Chr.10:1022637-1044201 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDI2-AS1 | NR_024628.1 | |||
| NR_024629.1 | ||||
| NR_027708.1 | ||||
| NR_027709.1 | ||||
| AF220183.1 | AAF67648.1 | |||
| AK314921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv5532 | Chr.10:1033559 - 1055760 on Build GRCh38 | Deletion |
|
| nsv1046220 | Chr.10:1018084 - 1143058 on Build GRCh38 | Gain |
|
| esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
| nsv951231 | Chr.10:1037361 - 1046360 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map