Assay Details
Target Gene Details
Entrez Gene ID: | 158835 |
Gene Name: | acyl-CoA wax alcohol acyltransferase 2 |
Gene Aliases: |
ARAT, DC4, DGAT2L4, MFAT, WS |
Location: |
Chr.X:70040542-70049970 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AWAT2 | NM_001002254.1 | NP_001002254.1 | ||
| XM_011530876.2 | XP_011529178.1 | |||
| AY605053.1 | AAT68764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3574029 | Chr.X:70039911 - 70050719 on Build GRCh38 | Loss |
|
| nsv6949 | Chr.X:70037194 - 70070943 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map