Assay Details
Target Gene Details
Entrez Gene ID: | 1271 |
Gene Name: | ciliary neurotrophic factor receptor |
Gene Aliases: |
- |
Location: |
Chr.9:34551432-34590386 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTFR | NM_001207011.1 | 10 | 1660 | NP_001193940.1 |
| NM_001842.4 | 9 | 1673 | NP_001833.1 | |
| NM_147164.2 | 10 | 1784 | NP_671693.1 | |
| XM_017014260.1 | 11 | 2000 | XP_016869749.1 | |
| XM_017014261.1 | 11 | 1973 | XP_016869750.1 | |
| XM_017014262.1 | 11 | 1917 | XP_016869751.1 | |
| XM_017014263.1 | 10 | 1758 | XP_016869752.1 | |
| XM_017014264.1 | 10 | 2302 | XP_016869753.1 | |
| XM_017014265.1 | 9 | 1618 | XP_016869754.1 | |
| AK127444.1 | 6 | 2765 | ||
| BC001492.2 | 9 | 1630 | AAH01492.1 | |
| BX364434.2 | 1 | 254 | ||
| DB311865.1 | 1 | 267 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion |
|
| nsv831543 | Chr.9:34431291 - 34615384 on Build GRCh38 | Loss |
|
| nsv519946 | Chr.9:34521869 - 34555001 on Build GRCh38 | Loss |
|
| nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map