Assay Details
Target Gene Details
Entrez Gene ID: | 128240 |
Gene Name: | NAD(P)HX epimerase |
Gene Aliases: |
AIBP, APOA1BP, YJEFN1 |
Location: |
Chr.1:156591762-156599818 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAXE | NM_144772.2 | NP_658985.2 | ||
| XM_017000319.1 | XP_016855808.1 | |||
| AJ315849.1 | CAC86580.1 | |||
| AK294835.1 | ||||
| AK298222.1 | ||||
| AK316083.1 | ||||
| BC056917.1 | AAH56917.1 | |||
| BC100931.1 | AAI00932.1 | |||
| BC100932.1 | AAI00933.1 | |||
| BC100933.1 | AAI00934.1 | |||
| BC100934.2 | AAI00935.1 | |||
| BG723681.1 | ||||
| BQ430064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map