Assay Details
Target Gene Details
Entrez Gene ID: | 10000 |
Gene Name: | AKT serine/threonine kinase 3 |
Gene Aliases: |
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2 |
Location: |
Chr.1:243488233-243851079 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKT3 | NM_001206729.1 | NP_001193658.1 | ||
| NM_005465.4 | 13 | 6795 | NP_005456.1 | |
| NM_181690.2 | NP_859029.1 | |||
| XM_005272994.4 | 14 | 7020 | XP_005273051.1 | |
| XM_005272995.2 | 14 | 6826 | XP_005273052.1 | |
| AL117525.1 | CAB55977.1 | |||
| AY005799.1 | AAF91073.1 | |||
| BC121154.2 | ||||
| U79271.1 | 1 | 898 |
Target Gene Details
Entrez Gene ID: | 10806 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP |
Location: |
Chr.1:243255328-243500092 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDCCAG8 | NM_006642.3 | 18 | 2532 | NP_006633.1 |
| XM_005273013.4 | 17 | 2364 | XP_005273070.1 | |
| XM_005273018.1 | 17 | 2371 | XP_005273075.1 | |
| XM_005273021.4 | 18 | 2304 | XP_005273078.1 | |
| XM_005273022.3 | 15 | 2153 | XP_005273079.1 | |
| XM_006711727.3 | 18 | 2418 | XP_006711790.1 | |
| XM_006711728.3 | 17 | 2289 | XP_006711791.1 | |
| XM_011544021.2 | 19 | 2514 | XP_011542323.1 | |
| XM_011544022.2 | 19 | 2589 | XP_011542324.1 | |
| XM_011544024.2 | 18 | 2387 | XP_011542326.1 | |
| XM_011544026.2 | 17 | 2277 | XP_011542328.1 | |
| XM_011544028.2 | 15 | 2052 | XP_011542330.1 | |
| XM_011544030.2 | 11 | 3065 | XP_011542332.1 | |
| XM_017000105.1 | 16 | 2282 | XP_016855594.1 | |
| XM_017000106.1 | 18 | 2500 | XP_016855595.1 | |
| XM_017000107.1 | 19 | 2416 | XP_016855596.1 | |
| XM_017000108.1 | 21 | 2963 | XP_016855597.1 | |
| AF039690.1 | 10 | 1466 | AAC18039.1 | |
| AK297105.1 | 14 | 1951 | ||
| AK299249.1 | 15 | 2124 | ||
| BC045832.1 | 7 | 971 | AAH45832.1 | |
| BX537630.1 | 1 | 3678 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833070 | Chr.1:243339809 - 243537501 on Build GRCh38 | Gain |
|
| nsv1013039 | Chr.1:243460621 - 243501269 on Build GRCh38 | Gain |
|
| nsv1010479 | Chr.1:243494728 - 243515087 on Build GRCh38 | Loss |
|
| dgv601n100 | Chr.1:243494646 - 243515087 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map