Assay Details
Target Gene Details
Entrez Gene ID: | 83416 |
Gene Name: | Fc receptor like 5 |
Gene Aliases: |
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820 |
Location: |
Chr.1:157513377-157552520 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL5 | NM_001195388.1 | 17 | 3307 | NP_001182317.1 |
| NM_031281.2 | 17 | 3317 | NP_112571.2 | |
| AF343664.1 | 17 | 3252 | AAK31327.1 | |
| AF369794.2 | 18 | 3317 | AAK50059.2 | |
| AK090423.1 | 9 | 2253 | BAC03404.1 | |
| AK090475.1 | 7 | 2436 | BAC03456.1 | |
| AL832301.1 | 2 | 547 | ||
| BC101066.2 | 17 | 3187 | AAI01067.1 | |
| BC101067.1 | 17 | 3187 | AAI01068.1 | |
| BC101069.2 | 17 | 3187 | AAI01070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831681 | Chr.1:157425048 - 157527722 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map