Assay Details
Target Gene Details
Entrez Gene ID: | 27086 |
Gene Name: | forkhead box P1 |
Gene Aliases: |
12CC4, HSPC215, MFH, QRF1, hFKH1B |
Location: |
Chr.3:70954714-71583989 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834727 | Chr.3:70884108 - 71058881 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map