Assay Details
Target Gene Details
Entrez Gene ID: | 8941 |
Gene Name: | cyclin dependent kinase 5 regulatory subunit 2 |
Gene Aliases: |
NCK5AI, P39, p39nck5ai |
Location: |
Chr.2:218959628-218962155 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK5R2 | NM_003936.4 | 1 | 2290 | NP_003927.1 |
| BC041771.1 | 1 | 2263 | AAH41771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584491 | Chr.2:218881839 - 219069626 on Build GRCh38 | Loss |
|
| nsv834542 | Chr.2:218841280 - 219013962 on Build GRCh38 | Gain |
|
| nsv3169 | Chr.2:218946755 - 218978274 on Build GRCh38 | Deletion |
|
| dgv7296n54 | Chr.2:218833281 - 218998934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map