Assay Details
Target Gene Details
Entrez Gene ID: | 51091 |
Gene Name: | Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase |
Gene Aliases: |
LP, PCH2D, SLA, SLA/LP |
Location: |
Chr.4:25120005-25160582 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPSECS | NM_016955.3 | 11 | 3354 | NP_058651.3 |
| XM_011513846.2 | 12 | 3193 | XP_011512148.1 | |
| XM_011513847.2 | 11 | 3460 | XP_011512149.1 | |
| XM_011513848.1 | 11 | 3597 | XP_011512150.1 | |
| XM_017008277.1 | 12 | 3573 | XP_016863766.1 | |
| XM_017008278.1 | 11 | 3328 | XP_016863767.1 | |
| AJ238617.1 | 11 | 3135 | CAB62209.1 | |
| BC117202.1 | 12 | 3198 | ||
| BC126213.1 | 11 | 3061 | ||
| BX648976.1 | 11 | 3215 | ||
| CN429794.1 | 1 | 532 |
More Information
Set Membership: |
|

Genomic Map