Assay Details
Target Gene Details
Entrez Gene ID: | 26005 |
Gene Name: | C2 calcium dependent domain containing 3 |
Gene Aliases: |
OFD14 |
Location: |
Chr.11:74012578-74171210 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 33 - Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2CD3 | NM_001286577.1 | NP_001273506.1 | ||
| NM_015531.5 | NP_056346.3 | |||
| XM_011544904.1 | XP_011543206.1 | |||
| XM_011544905.1 | XP_011543207.1 | |||
| XM_017017510.1 | XP_016872999.1 | |||
| XM_017017511.1 | XP_016873000.1 | |||
| XM_017017512.1 | XP_016873001.1 | |||
| XM_017017513.1 | XP_016873002.1 | |||
| XM_017017515.1 | XP_016873004.1 | |||
| XM_017017516.1 | XP_016873005.1 | |||
| AB231764.1 | BAE17137.1 | |||
| AI873280.1 | ||||
| AK125894.1 | BAC86334.1 | |||
| AL833903.1 | CAD38759.1 | |||
| BC035599.1 | AAH35599.1 | |||
| BC110508.2 | AAI10509.1 | |||
| CD369754.1 |
More Information
Set Membership: |
|

Genomic Map