Assay Details
Target Gene Details
Entrez Gene ID: | 56478 |
Gene Name: | eukaryotic translation initiation factor 4E nuclear import factor 1 |
Gene Aliases: |
4E-T, Clast4 |
Location: |
Chr.22:31436963-31489950 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4ENIF1 | NM_001164501.1 | NP_001157973.1 | ||
| NM_001164502.1 | NP_001157974.1 | |||
| NM_019843.3 | NP_062817.2 | |||
| XM_005261686.2 | XP_005261743.1 | |||
| XM_005261687.2 | XP_005261744.1 | |||
| XM_005261688.2 | XP_005261745.1 | |||
| XM_006724281.3 | XP_006724344.3 | |||
| XM_011530280.1 | XP_011528582.1 | |||
| XM_011530281.2 | XP_011528583.1 | |||
| XM_017028863.1 | XP_016884352.1 | |||
| XM_017028864.1 | XP_016884353.1 | |||
| AF240775.1 | AAF81693.1 | |||
| AK025254.1 | BAB15092.1 | |||
| AK074768.1 | BAC11194.1 | |||
| AK130061.1 | ||||
| AK299136.1 | ||||
| AK314636.1 | ||||
| BC032941.1 | AAH32941.1 | |||
| BC033028.1 | AAH33028.1 | |||
| CR456386.1 | CAG30272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834176 | Chr.22:31286440 - 31468715 on Build GRCh38 | Loss |
|
| nsv588894 | Chr.22:31437773 - 31581608 on Build GRCh38 | Loss |
|
| nsv1063634 | Chr.22:31401018 - 31525268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map