Assay Details
Target Gene Details
Entrez Gene ID: | 85397 |
Gene Name: | regulator of G-protein signaling 8 |
Gene Aliases: |
- |
Location: |
Chr.1:182641814-182753291 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS8 | NM_001102450.2 | 7 | 746 | NP_001095920.1 |
| NM_033345.3 | 6 | 617 | NP_203131.1 | |
| XM_005245555.3 | 7 | 760 | XP_005245612.1 | |
| XM_011510089.2 | 7 | 1657 | XP_011508391.1 | |
| XM_011510090.2 | 10 | 1487 | XP_011508392.1 | |
| XM_011510091.2 | 6 | 634 | XP_011508393.1 | |
| XM_017002631.1 | 10 | 1107 | XP_016858120.1 | |
| XM_017002632.1 | 8 | 938 | XP_016858121.1 | |
| XM_017002633.1 | 7 | 1136 | XP_016858122.1 | |
| XM_017002634.1 | 7 | 815 | XP_016858123.1 | |
| XM_017002635.1 | 8 | 1262 | XP_016858124.1 | |
| XM_017002636.1 | 6 | 744 | XP_016858125.1 | |
| XM_017002637.1 | 7 | 942 | XP_016858126.1 | |
| AF300649.1 | 5 | 455 | AAG18443.1 | |
| AI209102.1 | 1 | 171 | ||
| AK294660.1 | 7 | 713 | ||
| AK296514.1 | 7 | 706 | ||
| BC069677.1 | 7 | 695 | AAH69677.1 | |
| BC069718.1 | 6 | 567 | AAH69718.1 | |
| BC103865.1 | 7 | 722 | AAI03866.1 | |
| BC103866.1 | 7 | 722 | AAI03867.1 | |
| BC103867.1 | 7 | 722 | AAI03868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3810 | Chr.1:182630610 - 182661590 on Build GRCh38 | Deletion |
|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map