Assay Details
Target Gene Details
Entrez Gene ID: | 57678 |
Gene Name: | glycerol-3-phosphate acyltransferase, mitochondrial |
Gene Aliases: |
GPAT, GPAT1 |
Location: |
Chr.10:112149864-112215395 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPAM | NM_001244949.1 | 22 | 4249 | NP_001231878.1 |
| NM_020918.5 | 22 | 4245 | NP_065969.3 | |
| XM_005269998.1 | 22 | 4390 | XP_005270055.1 | |
| AB046780.1 | 16 | 3537 | BAB13386.1 | |
| AK025752.1 | 1 | 71 | ||
| AL833061.2 | 22 | 4231 | CAD89967.1 | |
| AL833093.2 | 22 | 4234 | CAD89932.1 | |
| BX647451.1 | 22 | 4213 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508608 | Chr.10:112030892 - 112158412 on Build GRCh38 | Deletion |
|
| nsv831989 | Chr.10:112112194 - 112301601 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73365159] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map