Assay Details
Target Gene Details
Entrez Gene ID: | 151126 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
ZNF533 |
Location: |
Chr.2:179441982-179864550 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385B | NM_001113397.1 | 8 | 2545 | NP_001106868.1 |
| NM_001113398.1 | 8 | 2405 | NP_001106869.1 | |
| NM_001282725.1 | 10 | 2499 | NP_001269654.1 | |
| NM_152520.4 | 10 | 3072 | NP_689733.3 | |
| NR_104234.1 | ||||
| XM_011510713.2 | 12 | 6213 | XP_011509015.1 | |
| XM_011510714.2 | 11 | 6163 | XP_011509016.1 | |
| XM_011510715.2 | 11 | 6116 | XP_011509017.1 | |
| XM_011510716.2 | 11 | 3324 | XP_011509018.1 | |
| XM_011510717.2 | 11 | 3321 | XP_011509019.1 | |
| XM_011510719.2 | 11 | 2584 | XP_011509021.1 | |
| XM_011510720.2 | 10 | 2444 | XP_011509022.1 | |
| XM_011510721.2 | 9 | 4737 | XP_011509023.1 | |
| XM_017003435.1 | 10 | 2753 | XP_016858924.1 | |
| XM_017003436.1 | 10 | 3231 | XP_016858925.1 | |
| XM_017003437.1 | 9 | 2388 | XP_016858926.1 | |
| XM_017003438.1 | 8 | 7871 | XP_016858927.1 | |
| XM_017003439.1 | 8 | 2379 | XP_016858928.1 | |
| XM_017003440.1 | 9 | 5908 | XP_016858929.1 | |
| AK057999.1 | 9 | 2288 | BAB71629.1 | |
| AK096822.1 | 8 | 2545 | BAC04870.1 | |
| BC048123.1 | AAH48123.1 | |||
| BC092423.1 | 10 | 2907 | AAH92423.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs111664903] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map