Assay Details
Target Gene Details
Entrez Gene ID: | 92935 |
Gene Name: | methionyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
COXPD25, MetRS, mtMetRS |
Location: |
Chr.2:197705304-197708390 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARS2 | NM_138395.3 | 1 | 2665 | NP_612404.1 |
| AB107013.1 | 1 | 2608 | BAC92749.1 | |
| AK098121.1 | 1 | 2600 | BAC05238.1 | |
| BC009115.1 | 1 | 2568 | AAH09115.1 | |
| BC040934.2 | 1 | 1643 | AAH40934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821871 | Chr.2:197707894 - 197708400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map