Assay Details
Target Gene Details
Entrez Gene ID: | 5985 |
Gene Name: | replication factor C subunit 5 |
Gene Aliases: |
RFC36 |
Location: |
Chr.12:118016701-118033568 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFC5 | XM_017019779.1 | 13 | 3165 | XP_016875268.1 |
Target Gene Details
Entrez Gene ID: | 55884 |
Gene Name: | WD repeat and SOCS box containing 2 |
Gene Aliases: |
SBA2 |
Location: |
Chr.12:118032687-118062430 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WSB2 | NM_001278557.1 | 9 | 2034 | NP_001265486.1 |
| NM_001278558.1 | 7 | 1757 | NP_001265487.1 | |
| NM_018639.4 | 9 | 2134 | NP_061109.1 | |
| XM_017019642.1 | 9 | 2036 | XP_016875131.1 | |
| AF038187.1 | 4 | 1263 | ||
| AF163324.1 | 8 | 1971 | AAF80478.1 | |
| AF229181.1 | 9 | 2073 | AAF71302.1 | |
| BC015887.2 | 9 | 2024 | AAH15887.1 | |
| CA443120.1 | 1 | 765 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
| nsv560388 | Chr.12:117953795 - 118044547 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map