Assay Details
Target Gene Details
Entrez Gene ID: | 80204 |
Gene Name: | F-box protein 11 |
Gene Aliases: |
FBX11, PRMT9, UBR6, UG063H01, VIT1 |
Location: |
Chr.2:47806920-47905793 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO11 | NM_001190274.1 | 19 | 2364 | NP_001177203.1 |
| NM_025133.4 | 19 | 2153 | NP_079409.3 | |
| XM_005264572.4 | 19 | 2358 | XP_005264629.1 | |
| XM_005264573.4 | 19 | 2311 | XP_005264630.1 | |
| XM_017005015.1 | 19 | 2311 | XP_016860504.1 | |
| XM_017005016.1 | 19 | 2177 | XP_016860505.1 | |
| XM_017005017.1 | 19 | 2177 | XP_016860506.1 | |
| AF264714.1 | 1 | 1299 | AAF76888.1 | |
| AK025477.1 | 4 | 356 | BAB15143.1 | |
| AK097734.1 | 8 | 901 | ||
| AK223592.1 | 19 | 2248 | BAD97312.1 | |
| AK292877.1 | 19 | 2203 | ||
| AK297444.1 | ||||
| AK304140.1 | 19 | 2068 | ||
| AK315108.1 | 19 | 2153 | ||
| AL117620.1 | 2 | 135 | CAB56019.1 | |
| AY827075.1 | 19 | 2291 | AAV87312.1 | |
| BC012728.2 | 5 | 437 | AAH12728.2 | |
| BC043258.1 | 19 | 2255 | AAH43258.2 | |
| BC130445.1 | 19 | 2089 | ||
| BC136480.1 | 19 | 2089 |
More Information
Set Membership: |
|

Genomic Map