Assay Details
Target Gene Details
Entrez Gene ID: | 10804 |
Gene Name: | gap junction protein beta 6 |
Gene Aliases: |
CX30, DFNA3, DFNA3B, DFNB1B, ECTD2, ED2, EDH, HED, HED2 |
Location: |
Chr.13:20221962-20232395 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB6 | NM_001110219.2 | 5 | 2069 | NP_001103689.1 |
| NM_001110220.2 | 4 | 1959 | NP_001103690.1 | |
| NM_001110221.2 | 3 | 1835 | NP_001103691.1 | |
| NM_006783.4 | 3 | 2001 | NP_006774.2 | |
| XM_017020357.1 | 4 | 2274 | XP_016875846.1 | |
| XM_017020358.1 | 5 | 1964 | XP_016875847.1 | |
| XM_017020359.1 | 5 | 1994 | XP_016875848.1 | |
| XM_017020360.1 | 4 | 1761 | XP_016875849.1 | |
| AK075247.1 | 5 | 1901 | ||
| BC038934.1 | 3 | 1696 | AAH38934.1 | |
| DB339720.1 | 1 | 63 | ||
| DB577868.1 | 1 | 408 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631424 | Chr.13:20190147 - 20280383 on Build GRCh38 | Gain |
|
| dgv3072n54 | Chr.13:20216603 - 20521581 on Build GRCh38 | Loss |
|
| esv3631426 | Chr.13:20219223 - 20261765 on Build GRCh38 | Gain |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
| esv3631421 | Chr.13:20145912 - 20223722 on Build GRCh38 | Gain |
|
| esv2751132 | Chr.13:20217943 - 20546665 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map