Assay Details
Target Gene Details
Entrez Gene ID: | 2515 |
Gene Name: | ADAM metallopeptidase domain 2 |
Gene Aliases: |
CRYN1, CRYN2, CT15, FTNB, PH-30b, PH30, PH30-beta |
Location: |
Chr.8:39743735-39838289 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM2 | NM_001278113.1 | NP_001265042.1 | ||
| NM_001278114.1 | NP_001265043.1 | |||
| NM_001464.4 | NP_001455.3 | |||
| XM_005273468.1 | XP_005273525.1 | |||
| XM_011544479.1 | XP_011542781.1 | |||
| XM_011544480.1 | XP_011542782.1 | |||
| AJ133005.1 | CAB40813.1 | |||
| AK301734.1 | ||||
| BC034957.2 | AAH34957.1 | |||
| BC064547.1 | AAH64547.1 | |||
| U38805.1 | AAD04206.1 | |||
| U52370.1 | AAC51110.1 | |||
| X99374.1 | CAA67753.1 |
More Information
Set Membership: |
|

Genomic Map