Assay Details
Target Gene Details
Entrez Gene ID: | 54979 |
Gene Name: | HRAS like suppressor 2 |
Gene Aliases: |
PLA1/2-2 |
Location: |
Chr.11:63552759-63565072 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRASLS2 | NM_017878.1 | 3 | 323 | NP_060348.1 |
| XM_011545120.2 | 4 | 775 | XP_011543422.1 | |
| AB453251.1 | 2 | 264 | ||
| AK000563.1 | 3 | 323 | BAA91256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626646 | Chr.11:63549774 - 63585192 on Build GRCh38 | Gain |
|
| esv3626647 | Chr.11:63558459 - 63582290 on Build GRCh38 | Gain |
|
| nsv352 | Chr.11:63555836 - 63589650 on Build GRCh38 | Insertion |
|
| nsv509417 | Chr.11:63485643 - 63569743 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map