Assay Details
Target Gene Details
Entrez Gene ID: | 401541 |
Gene Name: | centromere protein P |
Gene Aliases: |
CENP-P |
Location: |
Chr.9:92325457-92615164 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPP | NM_001012267.2 | NP_001012267.1 | ||
| NM_001286969.1 | NP_001273898.1 | |||
| XM_011518689.1 | XP_011516991.1 | |||
| AK092603.1 | ||||
| AK307435.1 | ||||
| BC071726.1 | AAH71726.1 | |||
| BX537851.1 | CAD97859.1 |
Target Gene Details
Entrez Gene ID: | 4969 |
Gene Name: | osteoglycin |
Gene Aliases: |
OG, OIF, SLRR3A |
Location: |
Chr.9:92383271-92404699 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGN | NM_014057.4 | 2 | 315 | NP_054776.1 |
| NM_024416.4 | NP_077727.3 | |||
| NM_033014.3 | 2 | 597 | NP_148935.1 | |
| AF086912.1 | 2 | 162 | AAP97142.1 | |
| AF100758.1 | 2 | 162 | AAD43022.1 | |
| AF202167.1 | 1 | 87 | AAF69109.1 | |
| AK075521.1 | 1 | 126 | ||
| AK129845.1 | 1 | 119 | ||
| AK289628.1 | 2 | 312 | ||
| AK295433.1 | ||||
| AL110267.1 | 1 | 104 | CAB53706.1 | |
| BC037273.2 | 1 | 103 | AAH37273.1 | |
| BC095443.1 | 1 | 119 | AAH95443.1 | |
| BG772245.1 | ||||
| BX646984.1 | 2 | 272 | ||
| CR533516.1 | CAG38547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053205 | Chr.9:92356913 - 92420330 on Build GRCh38 | Loss |
|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
| nsv1047337 | Chr.9:92343356 - 92585202 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map