Assay Details
Target Gene Details
Entrez Gene ID: | 5473 |
Gene Name: | pro-platelet basic protein |
Gene Aliases: |
B-TG1, Beta-TG, CTAP-III, CTAP3, CTAPIII, CXCL7, LA-PF4, LDGF, MDGF, NAP-2, PBP, SCYB7, TC1, TC2, TGB, TGB1, THBGB, THBGB1 |
Location: |
Chr.4:73986439-73988190 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPBP | NM_002704.3 | 1 | 78 | NP_002695.1 |
| AK312166.1 | 1 | 78 | ||
| BC028217.1 | 1 | 57 | AAH28217.1 | |
| BQ447470.1 | ||||
| BX443846.2 | 1 | 63 | ||
| CB112845.1 | 1 | 78 | ||
| CR456805.1 | CAG33086.1 | |||
| DA419455.1 | 1 | 78 | ||
| M54995.1 | AAA62836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006029 | Chr.4:73928205 - 74120947 on Build GRCh38 | Gain |
|
| esv3575747 | Chr.4:73914851 - 74124725 on Build GRCh38 | Gain |
|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map