Assay Details
Target Gene Details
Entrez Gene ID: | 55744 |
Gene Name: | cytochrome c oxidase assembly factor 1 homolog |
Gene Aliases: |
C7orf44, MITRAC15 |
Location: |
Chr.7:43608452-43729541 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COA1 | NR_135580.1 | |||
| NR_135581.1 | ||||
| NR_135582.1 | ||||
| NR_135583.1 | ||||
| XM_005249807.4 | 7 | 8228 | XP_005249864.1 | |
| XM_006715752.2 | 8 | 8196 | XP_006715815.1 | |
| XM_017012414.1 | 7 | 8155 | XP_016867903.1 | |
| XM_017012415.1 | 8 | 8257 | XP_016867904.1 | |
| XM_017012416.1 | XP_016867905.1 | |||
| XM_017012417.1 | 8 | 6659 | XP_016867906.1 | |
| XM_017012418.1 | 7 | 6529 | XP_016867907.1 | |
| XM_017012419.1 | XP_016867908.1 | |||
| XM_017012421.1 | 8 | 8379 | XP_016867910.1 | |
| XM_017012422.1 | 8 | 8268 | XP_016867911.1 | |
| XM_017012424.1 | 7 | 8171 | XP_016867913.1 | |
| XM_017012430.1 | 8 | 6631 | XP_016867919.1 | |
| XM_017012431.1 | XP_016867920.1 | |||
| XM_017012432.1 | XP_016867921.1 | |||
| AK023053.1 | BAB14379.1 | |||
| AK293043.1 | ||||
| BM918741.1 | ||||
| BX537586.1 | 1 | 1670 |
Target Gene Details
Entrez Gene ID: | 9263 |
Gene Name: | serine/threonine kinase 17a |
Gene Aliases: |
DRAK1 |
Location: |
Chr.7:43583093-43627379 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK17A | NM_004760.2 | 7 | 2524 | NP_004751.2 |
| XM_017012792.1 | 7 | 2336 | XP_016868281.1 | |
| AB011420.1 | 7 | 2490 | BAA34126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612996 | Chr.7:43537785 - 43656356 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map