Assay Details
Target Gene Details
Entrez Gene ID: | 26156 |
Gene Name: | ribosomal L1 domain containing 1 |
Gene Aliases: |
CSIG, L12, PBK1, UTP30 |
Location: |
Chr.16:11834196-11851585 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSL1D1 | NM_015659.2 | NP_056474.2 | ||
| AF083127.1 | AAF98239.1 | |||
| AJ007398.1 | CAA07491.1 | |||
| AK000822.1 | ||||
| AK295935.1 | ||||
| AL049999.1 | CAB43231.2 | |||
| AY154473.1 | AAN46298.1 | |||
| AY598331.1 | AAT06742.1 | |||
| BC016362.1 | ||||
| BC017899.1 | ||||
| BC019069.2 | AAH19069.2 | |||
| BC107782.1 | AAI07783.1 | |||
| BC112228.1 | AAI12229.1 | |||
| BC113699.1 | ||||
| BI856275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043909 | Chr.16:11746054 - 11913578 on Build GRCh38 | Gain |
|
| nsv1120318 | Chr.16:11824982 - 11855178 on Build GRCh38 | Tandem Duplication |
|
| nsv571458 | Chr.16:11831151 - 11993425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map