Assay Details
Target Gene Details
Entrez Gene ID: | 3899 |
Gene Name: | AF4/FMR2 family member 3 |
Gene Aliases: |
LAF4, MLLT2-like |
Location: |
Chr.2:99545861-100142583 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFF3 | NM_001025108.1 | 24 | 8042 | NP_001020279.1 |
| NM_002285.2 | 24 | 8059 | NP_002276.2 | |
| XM_005263943.3 | 23 | 7936 | XP_005264000.2 | |
| XM_005263945.3 | 15 | 7293 | XP_005264002.1 | |
| XM_011511169.2 | 24 | 8039 | XP_011509471.1 | |
| XM_011511170.2 | 25 | 8178 | XP_011509472.1 | |
| XM_011511171.2 | 24 | 7994 | XP_011509473.1 | |
| XM_011511172.2 | 23 | 7994 | XP_011509474.1 | |
| XM_011511173.2 | 24 | 8055 | XP_011509475.1 | |
| XM_011511174.2 | 24 | 7999 | XP_011509476.1 | |
| XM_011511175.2 | 26 | 8289 | XP_011509477.1 | |
| XM_011511176.2 | 23 | 7980 | XP_011509478.1 | |
| XM_011511177.2 | 24 | 8070 | XP_011509479.1 | |
| XM_011511178.2 | 14 | 6801 | XP_011509480.1 | |
| XM_011511179.2 | 14 | 6798 | XP_011509481.1 | |
| XM_017004085.1 | 15 | 7290 | XP_016859574.1 | |
| XM_017004086.1 | 15 | 7771 | XP_016859575.1 | |
| XM_017004087.1 | 15 | 7768 | XP_016859576.1 | |
| AK055117.1 | 1 | 1902 | ||
| BU740375.1 | 1 | 95 |
More Information
Set Membership: |
|

Genomic Map