Assay Details
Target Gene Details
Entrez Gene ID: | 28998 |
Gene Name: | mitochondrial ribosomal protein L13 |
Gene Aliases: |
L13, L13A, L13mt, RPL13, RPML13 |
Location: |
Chr.8:120395844-120445407 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL13 | NM_014078.5 | 7 | 955 | NP_054797.2 |
| AB049640.1 | 7 | 681 | BAB40845.1 | |
| AF112214.1 | 7 | 682 | AAF17202.1 | |
| AK123239.1 | 2 | 2216 | ||
| AK307322.1 | 6 | 559 | ||
| BC009190.2 | 7 | 686 | AAH09190.1 | |
| BC021744.2 | 7 | 934 | AAH21744.1 | |
| CN362173.1 | 3 | 269 |
More Information
Set Membership: |
|

Genomic Map