Assay Details
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP2 | XM_017004623.1 | XP_016860112.1 |
Target Gene Details
Entrez Gene ID: | 65124 |
Gene Name: | sosondowah ankyrin repeat domain family member C |
Gene Aliases: |
ANKRD57, C2orf26 |
Location: |
Chr.2:109614334-109618987 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOWAHC | NM_023016.3 | 1 | 2047 | NP_075392.2 |
| AK023346.1 | 1 | 327 | ||
| AK025523.1 | 1 | 2017 | BAB15161.1 | |
| BC036652.1 | 1 | 2071 | AAH36652.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829617 | Chr.2:109602280 - 109622435 on Build GRCh38 | Gain |
|
| esv2762780 | Chr.2:109610392 - 109704919 on Build GRCh38 | Gain |
|
| esv3591945 | Chr.2:109614455 - 109679968 on Build GRCh38 | Gain |
|
| dgv1162e212 | Chr.2:109504090 - 110630891 on Build GRCh38 | Gain |
|
| esv3591944 | Chr.2:109591382 - 109668039 on Build GRCh38 | Gain |
|
| dgv4037n100 | Chr.2:109591601 - 110226092 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map