Assay Details
Target Gene Details
Entrez Gene ID: | 55536 |
Gene Name: | cell division cycle associated 7 like |
Gene Aliases: |
JPO2, R1, RAM2 |
Location: |
Chr.7:21900899-21945924 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA7L | NM_001127370.2 | 11 | 2888 | NP_001120842.1 |
| NM_001127371.2 | 9 | 2696 | NP_001120843.1 | |
| NM_018719.4 | 10 | 2834 | NP_061189.2 | |
| AK022955.1 | 11 | 2378 | BAB14330.1 | |
| AK095965.1 | 11 | 2842 | ||
| AL359619.1 | 5 | 1960 | CAB94887.1 | |
| BC009352.2 | 10 | 2726 | AAH09352.2 | |
| BC014630.2 | 9 | 2678 | AAH14630.2 | |
| BC025242.1 | AAH25242.1 | |||
| BC032576.1 | AAH32576.1 | |||
| BC039823.1 | 10 | 2798 | AAH39823.1 | |
| BI493265.1 | 1 | 156 | ||
| BU753973.1 | 1 | 89 |
Target Gene Details
Entrez Gene ID: | 8701 |
Gene Name: | dynein axonemal heavy chain 11 |
Gene Aliases: |
CILD7, DNAHBL, DNAHC11, DNHBL, DPL11 |
Location: |
Chr.7:21543215-21901568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 81 - Exon 82 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAH11 | NM_001277115.1 | NP_001264044.1 | ||
| AJ320497.1 | CAC60121.1 | |||
| AK054657.1 | BAB70785.1 | |||
| AL529367.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830922 | Chr.7:21753768 - 21925057 on Build GRCh38 | Loss |
|
| nsv1027453 | Chr.7:21878856 - 21942830 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map