Assay Details
Target Gene Details
Entrez Gene ID: | 1108 |
Gene Name: | chromodomain helicase DNA binding protein 4 |
Gene Aliases: |
CHD-4, Mi-2b, Mi2-BETA |
Location: |
Chr.12:6570082-6607439 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD4 | NM_001273.3 | NP_001264.2 | ||
| NM_001297553.1 | NP_001284482.1 | |||
| XM_005253668.3 | XP_005253725.1 | |||
| XM_006718958.1 | 26 | 4358 | XP_006719021.1 | |
| XM_006718959.1 | XP_006719022.1 | |||
| XM_006718960.1 | XP_006719023.1 | |||
| XM_006718961.2 | XP_006719024.1 | |||
| XM_006718962.1 | XP_006719025.1 | |||
| XM_017018725.1 | XP_016874214.1 | |||
| XM_017018726.1 | XP_016874215.1 | |||
| XM_017018727.1 | XP_016874216.1 | |||
| XM_017018728.1 | XP_016874217.1 | |||
| XM_017018729.1 | XP_016874218.1 | |||
| XM_017018730.1 | XP_016874219.1 | |||
| XM_017018731.1 | XP_016874220.1 | |||
| XM_017018732.1 | XP_016874221.1 | |||
| XM_017018733.1 | XP_016874222.1 | |||
| XM_017018734.1 | XP_016874223.1 | |||
| AK128741.1 | ||||
| BC038596.1 | 26 | 4293 | AAH38596.1 | |
| CR936680.1 | ||||
| X86691.1 | CAA60384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509452 | Chr.12:6566935 - 6653627 on Build GRCh38 | Insertion |
|
| nsv826210 | Chr.12:6561863 - 6612310 on Build GRCh38 | Gain |
|
| nsv973982 | Chr.12:6578894 - 6587817 on Build GRCh38 | Duplication |
|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
| nsv826211 | Chr.12:6578422 - 6592697 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map